Anti-Borealin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-22832 NBP1-22832SS
NOVUNBP1-22832EA 554 EUR
NOVUNBP1-22832 NOVUNBP1-22832SS
Anti-Borealin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Borealin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR