Anti-RCAN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46853 NBP1-46853SS
NOVUNBP1-46853EA 554 EUR
NOVUNBP1-46853 NOVUNBP1-46853SS
Anti-RCAN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RCAN1 (regulator of calcineurin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR