Anti-N Cadherin Mouse Monoclonal Antibody [clone: 13A9]

Supplier: Novus Biologicals
NBP1-48309 NBP1-48309SS
NOVUNBP1-48309EA 638 EUR
NOVUNBP1-48309 NOVUNBP1-48309SS
Anti-N Cadherin Mouse Monoclonal Antibody [clone: 13A9]
Antibodies
Mouse Monoclonal N Cadherin Antibody (13A9). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:200

Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR