Anti-CXCR4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77560 NBP1-77560SS
NOVUNBP1-77560EA 638 EUR
NOVUNBP1-77560 NOVUNBP1-77560SS
Anti-CXCR4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR