Anti-GLI1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78259SS NBP1-78259
NOVUNBP1-78259SSEA 263 EUR
NOVUNBP1-78259SS NOVUNBP1-78259
Anti-GLI1 Rabbit Polyclonal Antibody
Antibodies
The GLI-1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to GLI-1. This antibody reacts with human, mouse. The GLI-1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: GLI-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR