Anti-B3GAT1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78982SS NBP1-78982
NOVUNBP1-78982SSEA 235 EUR
NOVUNBP1-78982SS NOVUNBP1-78982
Anti-B3GAT1 Rabbit Polyclonal Antibody
Antibodies
The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to beta-1,3-Glucuronyltransferase 1 / B3GAT1. This antibody reacts with human, mouse. The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:600

Type: Primary
Antigen: beta-1,3-Glucuronyltransferase 1/B3GAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR