Anti-GLP1R Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-97308 NBP1-97308SS
NOVUNBP1-97308EA 638 EUR
NOVUNBP1-97308 NOVUNBP1-97308SS
Anti-GLP1R Rabbit Polyclonal Antibody
Antibodies
The GLP-1R Antibody from Novus Biologicals is a rabbit polyclonal antibody to GLP-1R. This antibody reacts with human, mouse, rat. The GLP-1R Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: GLP-1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR