Anti-ATG12 Mouse Monoclonal Antibody [clone: 43CT73.3.5.5.4]

Supplier: United States Biological
USBI030788EA 587 EUR
USBI030788
Anti-ATG12 Mouse Monoclonal Antibody [clone: 43CT73.3.5.5.4]
Antibodies
Recommended dilution: ELISA: 1:10000; Western Blot: 1:200-2000; Immunochemistry: 1:50-200; Immunofluorescence:1:10-50

Type: Primary
Antigen: ATG12 (autophagy related 12)
Clonality: Monoclonal
Clone: 43CT73.3.5.5.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR