Anti-FOXD4L1 Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))

Supplier: US Biological
F9046-17-100 F9046-17-50
USBIF9046-17-100EA 270 EUR
USBIF9046-17-100 USBIF9046-17-50
Anti-FOXD4L1 Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))
Antibodies
Recommended dilution: ELISA: 1: 250000

Type: Primary
Antigen: FOXD4L1
Clonality: Polyclonal
Clone:
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR