Anti-Heat Shock Protein 70, Geodia cydonium Rabbit Polyclonal Antibody

Supplier: US Biological
H1834-35C-100 H1834-35C-20
USBIH1834-35C-100EA 838 EUR
USBIH1834-35C-100 USBIH1834-35C-20
Anti-Heat Shock Protein 70, Geodia cydonium Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:70000; Western Blot: 1:8000; Immunochemistry: 1:1000

Type: Primary
Antigen: Heat Shock Protein 70, Geodia cydonium
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR