Anti-Parathyroid Hormone Mouse Monoclonal Antibody [clone: B2-82
]

Supplier: US Biological
P3109-10H-10 P3109-10H-100
USBIP3109-10H-10EA 371 EUR
USBIP3109-10H-10 USBIP3109-10H-100
Anti-Parathyroid Hormone Mouse Monoclonal Antibody [clone: B2-82
]
Antibodies
Recommended dilution: ELISA: 1 ?g/ml; Immunochemistry: 2?g/ml; Radio Immunoassay: 25ng/ml

Type: Primary
Antigen: Parathyroid Hormone
Clonality: Monoclonal
Clone: B2-82

Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR