Anti-CaMKII Mouse Monoclonal Antibody [clone: 22B1]

Supplier: Cayman Chemical
10011438-100 10011438-25
CAYM10011438-100EA 641 EUR
CAYM10011438-100 CAYM10011438-25
Anti-CaMKII Mouse Monoclonal Antibody [clone: 22B1]
Antibodies
Type: Primary
Antigen: CaMKII
Clonality: Monoclonal
Clone: 22B1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR