Anti-FKBP52 Mouse Monoclonal Antibody [clone: Hi52c]

Supplier: Cayman Chemical
10011442-100 10011442-25
CAYM10011442-100EA 641 EUR
CAYM10011442-100 CAYM10011442-25
Anti-FKBP52 Mouse Monoclonal Antibody [clone: Hi52c]
Antibodies
Type: Primary
Antigen: FKBP52
Clonality: Monoclonal
Clone: Hi52c
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Dog, Hamster, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR