Anti-ANGPTL3 Rat Monoclonal Antibody [clone: Kairos3-1541]

Supplier: Cayman Chemical
10806-50 10806-100
CAYM10806-50EA 524 EUR
CAYM10806-50 CAYM10806-100
Anti-ANGPTL3 Rat Monoclonal Antibody [clone: Kairos3-1541]
Antibodies
Type: Primary
Antigen: ANGPTL3
Clonality: Monoclonal
Clone: Kairos3-1541
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR