Anti-Serpin A12 Mouse Monoclonal Antibody [clone: VP63]

Supplier: Cayman Chemical
10812-50 10812-100
CAYM10812-50EA 524 EUR
CAYM10812-50 CAYM10812-100
Anti-Serpin A12 Mouse Monoclonal Antibody [clone: VP63]
Antibodies
Type: Primary
Antigen: Serpin A12
Clonality: Monoclonal
Clone: VP63
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR