Anti-Ubiquitin Mouse Monoclonal Antibody [clone: 6C11-B3]

Supplier: Cayman Chemical
13723-50 13723-200
CAYM13723-50EA 380 EUR
CAYM13723-50 CAYM13723-200
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: 6C11-B3]
Antibodies
Type: Primary
Antigen: Ubiquitin
Clonality: Monoclonal
Clone: 6C11-B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR