Anti-Vtg Mouse Monoclonal Antibody [clone: JE-2A6]

Supplier: Cayman Chemical
170165-100 170165-500
CAYM170165-100EA 606 EUR
CAYM170165-100 CAYM170165-500
Anti-Vtg Mouse Monoclonal Antibody [clone: JE-2A6]
Antibodies
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: JE-2A6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR