N-(Hydroxymethyl)acrylamide for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.20675.0100 8.20675.1000
8.20675.0100EA 47.8 EUR
8.20675.0100 8.20675.1000
N-(Hydroxymethyl)acrylamide for synthesis, Sigma-Aldrich®
N-(Hydroxymethyl)acrylamide
Formula: C₄H₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00004597
CAS Number: 924-42-5

Order Now

Specification Test Results

Solubility 1880 g/l (20 °C)
Melting Point -10 °C
Boiling Point 100 °C (1013 hPa)
Vapor Pressure 31 hPa (25 °C)
Flash Point 93 °C
Density 1.08 g/cm3 (20 °C)
pH 6.0 - 7.0 (H2O, 20 °C)
Assay (ex N) 48 - 50%
Identity (IR) Passes test
Storage temperature 15 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR