Aluminum fluoride hydrate ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Scientific
Danger

10623.06 10623.22
10623.06EA 53.2 EUR
10623.06 10623.22
Aluminum fluoride hydrate ≥99.99% (metals basis), Puratronic®
Aluminum fluoride hydrate

Purity: 99.99% CAS No.: 32287-65-3 Molecular Formula: AlF3.xH2O Formula Weight: 83.98(anhy) Physical Form: -22 Mesh Powder UN No.: UN3260 MDL No.: MFCD00003426

Formula: AlF₃·xH₂O
MW: 83,98 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00003426
CAS Number: 32287-65-3
EINECS: 232-051-1
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR