trans-Diamminedichloropalladium(II) (≥49,9% Pd) ≥99.95% (metals basis), Premion®

Supplier: Thermo Scientific
Danger

Synonyms: trans-Dichlorodiammine palladium(II), Dichlorodiamminepalladium, trans-Diammine dichloropalladium

11037.02 11037.04 11037.09
11037.02EA 0 EUR
11037.02 11037.04 11037.09
trans-Diamminedichloropalladium(II) (≥49,9% Pd) ≥99.95% (metals basis), Premion®
trans-Diamminedichloropalladium(II)

Purity: 99.95% CAS No.: 14323-43-4 Molecular Formula: Pd(NH3)2Cl2 Formula Weight: 211.37 Physical Form: Crystalline powder Density: 2.5 Appearance: Yellow Solubility: Slightly soluble in water. Soluble in NH4OH. Insoluble in chloroform, acetone UN No.: UN3077 MDL No.: MFCD00011621

Formula: Cl₂H₆N₂Pd
MW: 211,39 g/mol
Density: 2,5
Storage Temperature: Ambient
MDL Number: MFCD00011621
CAS Number: 14323-43-4
EINECS: 238-269-3
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR