Dihydrogen hexahydroxyplatinate(IV) (≥61,0% Pt) ≥99.9% (metals basis)
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Hydrogen hexahydroxyplatinate(IV)
11052.02
11052.03
11052.06
11052.02EA
83.6
EUR
11052.02
11052.03
11052.06
Dihydrogen hexahydroxyplatinate(IV) (≥61,0% Pt) ≥99.9% (metals basis)
Dihydrogen hexahydroxyplatinate(IV)
Purity: 99.9%
CAS No.: 51850-20-5
Molecular Formula: H2Pt(OH)6
Formula Weight: 299.17
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Pale to dark yellow
Melting Point: dec.
MDL No.: MFCD00058744
Formula:
H₂Pt(OH)₆ MW: 299,14 g/mol Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00058744 CAS Number: 51850-20-5 EINECS: 257-471-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...