Iron(III) perchlorate hydrate, Reagent Grade
Supplier: Thermo Scientific
|
Danger
|
11647.22
11647.36
11647.22EA
65.5
EUR
11647.22
11647.36
Iron(III) perchlorate hydrate, Reagent Grade
Iron(III) perchlorate hydrate
CAS No.: 15201-61-3
Molecular Formula: Fe(ClO4)3.xH2O
Formula Weight: 354.19(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Yellow
UN No.: UN1481
MDL No.: MFCD00011000
Formula:
Cl₃FeH₂O₁₃ Storage Temperature: Ambient |
MDL Number:
MFCD00149705 CAS Number: 15201-61-3 EINECS: 236-908-0 UN: 1481 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...