Potassium hexachloroiridate(III) hydrate (≥32,8% Ir)
Supplier: Thermo Fisher Scientific
|
Warning
|
11887.03
11887.06
11887.14
11887.03EA
311
EUR
11887.03
11887.06
11887.14
Potassium hexachloroiridate(III) hydrate (≥32,8% Ir)
Potassium hexachloroiridate(III) hydrate
Molecular Formula: K3IrCl6.xH2O
Formula Weight: 522.22(anhy)
Physical Form: Crystalline
MDL No.: MFCD00038699
Formula:
K₃IrCl₆·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00038699 EINECS: 237-854-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...