Dihydrogen hexabromoplatinate(IV) nonahydrate (≥23,7% Pt) ≥99.9% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
12617.03
12617.06
12617.03EA
83.9
EUR
12617.03
12617.06
Dihydrogen hexabromoplatinate(IV) nonahydrate (≥23,7% Pt) ≥99.9% (metals basis)
Dihydrogen hexabromoplatinate(IV) nonahydrate
Purity: 99.9%
CAS No.: 20596-34-3
Molecular Formula: H2Br6Pt.9H2O
Formula Weight: 838.67 (676.56anhy)
Physical Form: Crystalline
Appearance: Red-brown
UN No.: UN3260
MDL No.: MFCD00049645
Formula:
H₂Br₆Pt·₉H₂O MW: 838,67 g/mol (676,56 anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00049645 CAS Number: 20596-34-3 EINECS: 243-903-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...