Cesium propionate, complex with propionic acid

Supplier: Thermo Fisher Scientific
Warning

18865.04 18865.18
18865.04EA 0 EUR
18865.04 18865.18
Cesium propionate, complex with propionic acid
Cesium propionate, complex with propionic acid

CAS No.: 38869-24-8 Molecular Formula: CsO2CC2H5.CH3CH2CO2H Formula Weight: 280.06 Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00014025

Formula: CsO₂CC₂H₅·CH₃CH₂CO₂H
Storage Temperature: Ambient
MDL Number: MFCD00014025
CAS Number: 38869-24-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR