Zinc p-toluenesulphonate hydrate

Supplier: Thermo Scientific
Warning

Synonyms: Zinc p-toluenesulphonate xhydrate

20899.14 20899.22
20899.14EA 63.4 EUR
20899.14 20899.22
Zinc p-toluenesulphonate hydrate
Zinc p-toluenesulphonate hydrate

CAS No.: 123334-05-4 Molecular Formula: C14H14O6S2Zn.xH2O Formula Weight: 407.76 Physical Form: Powder MDL No.: MFCD00064811

Formula: C₁₄H₁₆O₇S₂Zn
MW: 425,775 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064811
CAS Number: 123334-05-4
EINECS: 236-576-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR