Barium metaphosphate ≥90%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Bariummetaphosphate
22641.22
22641.36
22641.22EA
0
EUR
22641.22
22641.36
Barium metaphosphate ≥90%
Barium metaphosphate
Purity: 90+%
CAS No.: 13762-83-9
Molecular Formula: Ba(PO3)2
Formula Weight: 295.25
Physical Form: Powder
Odour: Odourless
Melting Point: 1560°
Solubility: Insoluble in water. Soluble in acids by a slow dissolution
UN No.: UN1564
MDL No.: MFCD00061417
Formula:
Ba(PO₃)₂ Melting Pt: 1560 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061417 CAS Number: 13762-83-9 EINECS: 237-362-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...