Ammonium O,O'-diethyl dithiophosphate ≥95%
Supplier: Thermo Fisher Scientific
30139.06
30139.14
30139.06EA
57
EUR
30139.06
30139.14
Ammonium O,O'-diethyl dithiophosphate ≥95%
Ammonium O,O'-diethyl dithiophosphate
Purity: 95%
CAS No.: 1068-22-0
Molecular Formula: C4H14NO2PS2
Formula Weight: 203.25
Physical Form: Crystalline
Appearance: White
Odour: Unpleasant
Melting Point: 165-175°
MDL No.: MFCD00012635
Formula:
C₄H₁₄NO₂PS₂ Melting Pt: 165…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012635 CAS Number: 1068-22-0 EINECS: 213-942-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...