1,3-Bis(diphenylphosphino)propanenickel(II) chloride

Supplier: Thermo Scientific
Warning

Synonyms: [1,3-Bis(diphenylphosphino)propane]nickel (II) chloride, Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), 1,3-Bis(diphenylphosphino)propane nickel(II)chloride, Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II), [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), [1,3-Bis(diphenylphosphino)propane]nickel(II) dichloride

30167.04 30167.09
30167.04EA 60.2 EUR
30167.04 30167.09
1,3-Bis(diphenylphosphino)propanenickel(II) chloride
1,3-Bis(diphenylphosphino)propanenickel(II) chloride

CAS No.: 15629-92-2 Molecular Formula: C27H26Cl2NiP2 Formula Weight: 542.03 Physical Form: Powder Melting Point: 213° dec. MDL No.: MFCD00015318

Formula: C₂₇H₂₆Cl₂NiP₂
MW: 542,042 g/mol
Melting Pt: 213 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00015318
CAS Number: 15629-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR