Dimethylphosphinic chloride ≥97%
Supplier: Thermo Scientific
|
Danger
|
30292.02
30292.04
30292.02EA
135
EUR
30292.02
30292.04
Dimethylphosphinic chloride ≥97%
Dimethylphosphinic chloride
Purity: 97+%
CAS No.: 1111-92-8
Molecular Formula: (CH3)2P(O)Cl
Formula Weight: 112.51
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 68°
Boiling Point: 204°
UN No.: UN3261
MDL No.: MFCD00002078
Formula:
C₂H₆ClOP MW: 112,493 g/mol Boiling Pt: 204 °C Melting Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002078 CAS Number: 1111-92-8 EINECS: 214-187-3 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...