Dodecylbenzene mixture of isomers

Supplier: Thermo Scientific

Synonyms: n-Dodecylbenzene, 1-Phenyldodecane

31362.30 31362.A1
31362.30EA 162 EUR
31362.30 31362.A1
Dodecylbenzene mixture of isomers
Dodecylbenzene

CAS No.: 123-01-3 Molecular Formula: C6H5C12H25 Formula Weight: 246.44 Physical Form: Liquid Density: 0.87 Refractive Index: 1.49 Odour: Specific type Melting Point: -3° Boiling Point: 179-180°/13mm MDL No.: MFCD00008974

Formula: C₁₈H₃₀
MW: 246,44 g/mol
Boiling Pt: 179…180 °C (13 mmHg)
Melting Pt: –3 °C
Density: 0,87
Storage Temperature: Ambient
MDL Number: MFCD00008974
CAS Number: 123-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR