Insulating beads, ceramic, AL23
Supplier: Thermo Scientific
32911.KX 32915.KX
32911.KXEA 0 EUR
32911.KX 32915.KX
Insulating beads, ceramic, AL23
Beads Ceramic Beads
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.