Bis[2-carboxyethylgermanium(IV)] sesquioxide ≥99.7%
Supplier: Thermo Fisher Scientific
Warning
|
36758.06
36758.14
36758.22
36758.06EA
88.9
EUR
36758.06
36758.14
36758.22
Bis[2-carboxyethylgermanium(IV)] sesquioxide ≥99.7%
Bis[2-carboxyethylgermanium(IV)] sesquioxide
Purity: 99.7%
CAS No.: 12758-40-6
Molecular Formula: C6H10Ge2O7
Formula Weight: 339.25
Physical Form: Powder
Odour: Odourless
Melting Point: 320° dec.
MDL No.: MFCD00049319
Formula:
C₆H₁₀Ge₂O₇ MW: 339.40 g/mol Melting Pt: 320 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00049319 CAS Number: 12758-40-6 EINECS: 235-800-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...