Lithium niobium ethoxide 5% (w/v) in ethanol , 99% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
39155.AC
39155.ACEA
103
EUR
39155.AC
Lithium niobium ethoxide 5% (w/v) in ethanol , 99% (metals basis)
Lithium niobium ethoxide
Purity: 99+%
CAS No.: 86745-52-0
Molecular Formula: LiNb(OC2H5)6
Formula Weight: 370.21
Flash Point: 17°(63°F)
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Yellow
Odour: Alcohol-like
UN No.: UN1170
MDL No.: MFCD00210625
Formula:
LiNb(OC₂H₅)₆ MW: 370,21 g/mol Flash Pt: 17 °C (63 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00210625 CAS Number: 86745-52-0 UN: 1170 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...