Bis(cyclopentadienyl)molybdenum dichloride ≥99%

Supplier: Thermo Scientific
Danger


Packaged under argon.

39393.02 39393.04
39393.02EA 146 EUR
39393.02 39393.04
Bis(cyclopentadienyl)molybdenum dichloride ≥99%
Bis(cyclopentadienyl)molybdenum dichloride

Purity: 99% CAS No.: 12184-22-4 Molecular Formula: C10H10Cl2Mo Formula Weight: 297.04 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive UN No.: UN3261 MDL No.: MFCD00009948

Formula: C₁₀H₁₀Cl₂Mo
MW: 297,032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009948
CAS Number: 12184-22-4
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR