Samarium(III) hydroxide hydrate ≥99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific
|
Danger
|
39512.06
39512.14
39512.06EA
80.4
EUR
39512.06
39512.14
Samarium(III) hydroxide hydrate ≥99% (REO, rare earth oxide basis)
Samarium(III) hydroxide hydrate
Purity: 99%
CAS No.: 20403-06-9
Molecular Formula: Sm(OH)3.xH2O
Formula Weight: 201.37(anhy)
Physical Form: Lump/Powder
UN No.: UN3262
MDL No.: MFCD00050108
Formula:
Sm(OH)₃·xH₂O MW: 201,37 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00050108 CAS Number: 20403-06-9 EINECS: 243-787-8 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...