Cesium metasilicate ≥99%
Supplier: Thermo Scientific
|
Warning
|
40208.14
40208.22
40208.14EA
222
EUR
40208.14
40208.22
Cesium metasilicate ≥99%
Cesium metasilicate
Purity: 99%
CAS No.: 15586-77-3
Molecular Formula: Cs2SiO3
Formula Weight: 341.89
Physical Form: Powder
Odour: Odourless
MDL No.: MFCD00798519
Formula:
Cs₂O₃Si MW: 341,893 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798519 CAS Number: 15586-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...