Cyclopentadienylmolybdenum(V) tetrachloride ≥95%
Supplier: Thermo Scientific
|
Danger
|
40522.06
40522.MD
40522.06EA
603
EUR
40522.06
40522.MD
Cyclopentadienylmolybdenum(V) tetrachloride ≥95%
Cyclopentadienylmolybdenum(V) tetrachloride
Purity: 95%
CAS No.: 62927-99-5
Molecular Formula: C5H5Cl4Mo
Formula Weight: 302.85
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Red-purple
UN No.: UN3261
MDL No.: MFCD00269826
Formula:
C₅H₅Cl₄Mo MW: 302,845 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00269826 CAS Number: 62927-99-5 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...