Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.05 mm (0.002 in) Type 304
Supplier: Thermo Fisher Scientific
41582.FF
41582.FI
41582.GH
41582.FFEA
41.5
EUR
41582.FF
41582.FI
41582.GH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.05 mm (0.002 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...