Cobalt titanate (<0,15% Ni) ≥99.8% (metals basis excluding Ni)
Supplier: Thermo Scientific
|
Danger
|
41670.18
41670.30
41670.18EA
122
EUR
41670.18
41670.30
Cobalt titanate (<0,15% Ni) ≥99.8% (metals basis excluding Ni)
Cobalt titanate
Purity: 99.8%
CAS No.: 12017-01-5
Molecular Formula: CoTiO3, may contain small amounts of other phases
Formula Weight: 154.83
Physical Form: -325 Mesh Powder
MDL No.: MFCD00016022
Formula:
CoO₃Ti Storage Temperature: Ambient |
MDL Number:
MFCD00016022 CAS Number: 12017-01-5 EINECS: 234-615-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...