Humic acid sodium salt 50 - 60% (as humic acid), tech.

Supplier: Thermo Fisher Scientific

Synonyms: Sodium bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate

41748.22 41748.36
41748.22EA 72.9 EUR
41748.22 41748.36
Humic acid sodium salt 50 - 60% (as humic acid), tech.
Humic acid sodium salt

Purity: 50-60% CAS No.: 68131-04-4 Physical Form: Powder Appearance: Black Melting Point: 300° MDL No.: MFCD00135560

Formula: C₉H₈Na₂O₄
MW: 226.139 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00135560
CAS Number: 68131-04-4
EINECS: 268-608-0
Merck Index: 14,4752

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR