Polyimide, film, Thickness: 0.008 mm

Supplier: Thermo Fisher Scientific

41896.GH 41896.GT 41896.HB
41896.GHEA 261 EUR
41896.GH 41896.GT 41896.HB
Polyimide, film, Thickness: 0.008 mm
Polyimide

CAS No.: 62929-02-6 Physical Form: ≈0.1136g/100x100mm MDL No.: MFCD00677744

Formula: C₃₅H₂₈N₂O₇
Storage Temperature: Ambient
MDL Number: MFCD01868102
CAS Number: 62929-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR