Europium(III) phosphate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific
Warning

41989.03 41989.06
41989.03EA 59.1 EUR
41989.03 41989.06
Europium(III) phosphate hydrate ≥99.9% (REO, rare earth oxide basis)
Europium(III) phosphate hydrate

Purity: 99.9% CAS No.: 14913-20-3 Molecular Formula: EuO4P.xH2O Formula Weight: 246.93(anhy) Physical Form: Chunks/powder Sensitivity: Hygroscopic MDL No.: MFCD05664261

Formula: EuO₄P·xH₂O
MW: 246,93 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD05664261
CAS Number: 14913-20-3
EINECS: 236-901-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR