Palladium : Platinum (8:2) on activated carbon wetted with ca. 50% water, powder
Supplier: Thermo Scientific
42208.04
42208.09
42208.04EA
73.4
EUR
42208.04
42208.09
Palladium : Platinum (8:2) on activated carbon wetted with ca. 50% water, powder
Palladium : Platinum
Uses: Selective hydrogenation of nitrates to hydroxylamines, reduction of nitrogen oxides to N2 with H2 used in presence of CO and CO2
Sold on dry weight basis. Unit weight excludes water weight
CAS No.: 7440-05-3
MDL No.: MFCD01074898
Applications: Selective hydrogenation of nitrates to hydroxylamines, reduction of nitrogen oxides to N2 with H2 in the presence of CO and CO2
Formula:
Pd:Pt Storage Temperature: Ambient |
MDL Number:
MFCD03613605 CAS Number: 7440-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...