Gadolinium(III) isopropoxide 5% (w/v) in toluene/isopropanol (3:2) , 99.9% (metals basis)

Supplier: Thermo Scientific
Danger

Synonyms: Gadolinium(III) i-propoxide

42277.AC 42277.AE
42277.ACEA 0 EUR
42277.AC 42277.AE
Gadolinium(III) isopropoxide 5% (w/v) in toluene/isopropanol (3:2) , 99.9% (metals basis)
Gadolinium(III) isopropoxide

Purity: 99.9% CAS No.: 14532-05-9 Molecular Formula: C9H21GdO3 Formula Weight: 334.51 Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Pale yellow Odour: Aromatic UN No.: UN1993 MDL No.: MFCD00144381


Applications: Precursor for Gd2O3 films using Sol-Gel process

Formula: C₉H₂₁GdO₃
MW: 334,514 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144381
CAS Number: 14532-05-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR