Lanthanum(III) phosphate hydrate
Supplier: Thermo Fisher Scientific
43125.14
43125.22
43125.14EA
94.3
EUR
43125.14
43125.22
Lanthanum(III) phosphate hydrate
Lanthanum(III) phosphate hydrate
CAS No.: 14913-14-5
Molecular Formula: LaPO4.xH2O
Formula Weight: 233.88(anhy)
Physical Form: Lump
Odour: Odourless
MDL No.: MFCD00150248
Formula:
LaPO₄.xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150248 CAS Number: 14913-14-5 EINECS: 237-419-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...