6-Methyl-4-trifluoromethyl-2(1H)-pyridone ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one
43467.03
43467.06
43467.03EA
110
EUR
43467.03
43467.06
6-Methyl-4-trifluoromethyl-2(1H)-pyridone ≥97%
6-Methyl-4-trifluoromethyl-2(1H)-pyridone
Purity: 97%
CAS No.: 22123-19-9
Molecular Formula: C7H6F3NO
Formula Weight: 177.13
Physical Form: Powder
Melting Point: 129-130°
MDL No.: MFCD00042479
Formula:
C₇H₆F₃NO MW: 177,126 g/mol Melting Pt: 129…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042479 CAS Number: 22123-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...