Trimethyl(pentamethylcyclopentadienyl)titanium(IV) ≥97%
Supplier: Thermo Scientific
|
Danger
|
ampouled under argon
43759.02
43759.04
43759.02EA
285
EUR
43759.02
43759.04
Trimethyl(pentamethylcyclopentadienyl)titanium(IV) ≥97%
Trimethyl(pentamethylcyclopentadienyl)titanium(IV)
Purity: 97%
CAS No.: 107333-47-1
Molecular Formula: C13H24Ti
Formula Weight: 228.22
Physical Form: Crystalline, Ampouled under argon
Sensitivity: Light, Air and Moisture Sensitive
UN No.: UN1325
MDL No.: MFCD00269851
Formula:
C₁₃H₂₄Ti Storage Temperature: Ambient |
MDL Number:
MFCD00269851 CAS Number: 107333-47-1 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...