3-Methyl-1-phenylindolin-2-one ≥97%
Supplier: Thermo Scientific
43808.03
43808.06
43808.03EA
126
EUR
43808.03
43808.06
3-Methyl-1-phenylindolin-2-one ≥97%
3-Methyl-1-phenylindolin-2-one
Purity: 97%
CAS No.: 23210-22-2
Molecular Formula: C15H13NO
Formula Weight: 223.27
Physical Form: Powder
MDL No.: MFCD00798609
Formula:
C₁₅H₁₃NO MW: 223,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798609 CAS Number: 23210-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...