3-Methyl-3-trimethylsiloxy-1-butyne ≥97%
Supplier: Thermo Scientific
|
Danger
|
43922.09
43922.18
43922.09EA
115
EUR
43922.09
43922.18
3-Methyl-3-trimethylsiloxy-1-butyne ≥97%
3-Methyl-3-trimethylsiloxy-1-butyne
Purity: 97%
CAS No.: 17869-77-1
Molecular Formula: HC≡CC[OSi(CH3)3](CH3)CH3
Formula Weight: 156.30
Physical Form: Liquid
Refractive Index: 1.4052
Odour: Characteristic
Boiling Point: 52°/75mm
UN No.: UN1993
MDL No.: MFCD00053867
Formula:
HC ΞCC[OSi(CH₃)₃](CH₃)CH₃ MW: 156,30 g/mol Boiling Pt: 52 °C (75 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00053867 CAS Number: 17869-77-1 EINECS: 241-823-7 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...